Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC34A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120232
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120232 Supplier Invitrogen™ Supplier No. PA5120232
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, A-549, U-87MG, Mouse kidney, Rat lung. The target is usually found in the following locations: Membrane, Multi-pass membrane protein. Immunogen sequence: QSRSPGHLPK WLQTWDFLPR WMHSLKPLDH LITRATLCCA RPEPRSPPLP PRVFLEELPP ATPSPRLALP AHHNATRL.

Renal tubular reabsorption of phosphate is critical to the maintenance of phosphate homeostasis in mammals. The brush-border membrane Na+/Pi cotransport systems in proximal tubules play a major role in this process. The renal Na(+)/P(i) cotransporter NPT2 is expressed in the brush border membrane (BBM) of proximal tubular cells. NPT2 gene expression is crucial for PTH effects on renal P(i) handling. However, renal expression of the sodium/phosphate cotransporter gene, NPT2, is not required for regulation of renal 1 alpha-hydroxylase by phosphate. NPT2 is an integral membrane protein expressed in kidney and lung. The gene encoding human NPT1 maps to chromosome 6q21.3-p23, while the gene encoding human NPT2 maps to chromosome 5q35.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC34A1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SLC34A1
Gene Accession No. Q06495, Q06496, Q60825
Gene Alias FRTS2; HCINF2; Na(+)/Pi cotransporter 2A; Na(+)-dependent phosphate cotransporter 2A; Na/Pi cotransporter; Na+-phosphate cotransporter type II; NaPi-2; NaPi2A; naPi-2a; NAPI-3; naPi-7; NaPi-IIa; NPHLOP1; NPT2; Npt2a; NPTIIa; renal Na+/Pi transporter; renal sodium-dependent phosphate transporter; SLC11; Slc17a2; Slc34a1; sodium/phosphate co-transporter; sodium/phosphate cotransporter 2A; Sodium-dependent phosphate transport protein 2A; sodium-phosphate transport protein 2A; solute carrier family 17 (sodium phosphate), member 2; solute carrier family 17 (sodium/hydrogen exchanger) member 2; solute carrier family 17 (sodium/hydrogen exchanger), member 2; solute carrier family 34 (sodium phosphate), member 1; solute carrier family 34 (type II sodium/phosphate cotransporter), member 1; solute carrier family 34 member 1; solute carrier family 34, member 1; type IIa Na+/Pi-cotransporter
Gene Symbols SLC34A1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20505, 25548, 6569
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.