Learn More
SLC34A1 Polyclonal Antibody

Description
SLC34A1 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | SLC34A1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | SLC34A1 |
| Gene Accession No. | Q06496 |
| Gene Alias | FRTS2; HCINF2; Na(+)/Pi cotransporter 2A; Na(+)-dependent phosphate cotransporter 2A; Na/Pi cotransporter; Na+-phosphate cotransporter type II; NaPi-2; NaPi2A; naPi-2a; NAPI-3; naPi-7; NaPi-IIa; NPHLOP1; NPT2; Npt2a; NPTIIa; renal Na+/Pi transporter; renal sodium-dependent phosphate transporter; SLC11; Slc17a2; Slc34a1; sodium/phosphate co-transporter; sodium/phosphate cotransporter 2A; Sodium-dependent phosphate transport protein 2A; sodium-phosphate transport protein 2A; solute carrier family 17 (sodium phosphate), member 2; solute carrier family 17 (sodium/hydrogen exchanger) member 2; solute carrier family 17 (sodium/hydrogen exchanger), member 2; solute carrier family 34 (sodium phosphate), member 1; solute carrier family 34 (type II sodium/phosphate cotransporter), member 1; solute carrier family 34 member 1; solute carrier family 34, member 1; type IIa Na+/Pi-cotransporter |
| Gene Symbols | SLC34A1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.