Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC34A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100689
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100689 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100689 Supplier Invitrogen™ Supplier No. PA5100689
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC34A2.

SLC34A2 encodes a protein that is a pH-sensitive sodium-dependent phosphate transporter. Phosphate uptake is increased at lower pH. Defects in this gene are a cause of pulmonary alveolar microlithiasis. Three transcript variants encoding two different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC34A2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC34A2
Gene Accession No. O95436, Q9DBP0
Gene Alias AA536683; D5Ertd227e; Na(+)/Pi cotransporter 2B; Na(+)-dependent phosphate cotransporter 2B; NAPI IIb; NaPi-2b; NaPi3b; NAPI-3B; NAPI-IIb; Npt2b; NPTIIb; rNaPi IIb; Slc34a2; Sodium/phosphate cotransporter 2B; Sodium-dependent phosphate transport protein 2B; sodium-phosphate transport protein 2B; solute carrier family 34 (sodium phosphate), member 2; solute carrier family 34 (type II sodium/phosphate contransporter), member 2; solute carrier family 34 (type II sodium/phosphate cotransporter), member 2; solute carrier family 34 member 2; type II sodium-dependent phosphate transporter 3b; type IIb Na/Picotransporter; type IIb sodium-phosphate transporter
Gene Symbols SLC34A2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC34A2(Accession O95436), corresponding to amino acid residues G8-T58.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10568, 20531
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.