Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC34A3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30636650UL
Description
SLC34A3 Polyclonal antibody specifically detects SLC34A3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
SLC34A3 | |
Polyclonal | |
Immunohistochemistry-Paraffin 1:200-1:1000 | |
Na(+)/Pi cotransporter 2C, Na(+)-dependent phosphate cotransporter 2C, naPi-2c, sodium-phosphate transport protein 2C, solute carrier family 34 (sodium phosphate), member 3 | |
Produced in rabbits immunized with E. coli-derived Human SLC34A3 fragment. | |
50 μg | |
Cancer, Endocrinology, Signal Transduction | |
142680 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH7.0 | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction