Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC34A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554883
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554883 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554883 Supplier Invitrogen™ Supplier No. PA554883
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPSSLPGSQV PHPTLDAVDL VEKTLRNEGT SSSAPVLEEG DTDPWTLPQL KDTSQPWKEL RVAGRLRRVA GSVLKACG Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 60%.

This gene encodes a member of SLC34A transporter family of proteins, and is expressed primarily in the kidney. It is involved in transporting phosphate into cells via sodium cotransport in the renal brush border membrane, and contributes to the maintenance of inorganic phosphate concentration in the kidney. Mutations in this gene are associated with hereditary hypophosphatemic rickets with hypercalciuria. Alternatively spliced transcript variants varying in the 5' UTR have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC34A3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC34A3
Gene Accession No. Q8N130
Gene Alias AI649385; HHRH; Na(+)/Pi cotransporter 2C; Na(+)-dependent phosphate cotransporter 2C; Na+/Pi-cotransporter type IIc; naPi-2c; Npt2c; Npt2c-v1; NptIIc; Slc34a3; Sodium/inorganic phosphate cotransporter IIC; sodium/phosphate cotransporter 2C; Sodium-dependent phosphate transport protein 2C; sodium-dependent phosphate transport protein 2C; LOW QUALITY PROTEIN: sodium-dependent phosphate transport protein 2C; sodium-phosphate cotransporter IIC; sodium-phosphate transport protein 2C; solute carrier family 34 (sodium phosphate), member 3; solute carrier family 34 (type II sodium/phosphate contransporter), member 3; solute carrier family 34 (type II sodium/phosphate cotransporter), member 3; solute carrier family 34 member 3; type IIc Na+/Pi cotransporter; type IIc Na+/Pi-cotransporter; type IIc NaPi-cotransporter-v1
Gene Symbols SLC34A3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC34A3. Recombinant protein control fragment (Product #RP-93101).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 142680
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.