Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC34A3 Polyclonal Antibody

Catalog No. PIPA595706
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595706 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595706 Supplier Invitrogen™ Supplier No. PA595706

Rabbit Polyclonal Antibody

Positive Samples: Mouse brain, Mouse kidney, Rat brain; Cellular Location: Membrane, Multi-pass membrane protein Immunogen sequence: MAAAAAAGSG TPREEEGPAG EAAASQPQAP TSVPGARLSR LPLARVKALV KADPDVTLAG QEAIFILARA AELFVETIAK DAYCCAQQGK RKTLQRRDLD NAIEAVDEFA FLEGTLD.

This gene encodes a member of SLC34A transporter family of proteins, and is expressed primarily in the kidney. It is involved in transporting phosphate into cells via sodium cotransport in the renal brush border membrane, and contributes to the maintenance of inorganic phosphate concentration in the kidney. Mutations in this gene are associated with hereditary hypophosphatemic rickets with hypercalciuria. Alternatively spliced transcript variants varying in the 5' UTR have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC34A3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SLC34A3
Gene Accession No. Q80SU6, Q8K4R8, Q8N130
Gene Alias AI649385; HHRH; Na(+)/Pi cotransporter 2C; Na(+)-dependent phosphate cotransporter 2C; Na+/Pi-cotransporter type IIc; naPi-2c; Npt2c; Npt2c-v1; NptIIc; Slc34a3; Sodium/inorganic phosphate cotransporter IIC; sodium/phosphate cotransporter 2C; Sodium-dependent phosphate transport protein 2C; sodium-dependent phosphate transport protein 2C; LOW QUALITY PROTEIN: sodium-dependent phosphate transport protein 2C; sodium-phosphate cotransporter IIC; sodium-phosphate transport protein 2C; solute carrier family 34 (sodium phosphate), member 3; solute carrier family 34 (type II sodium/phosphate contransporter), member 3; solute carrier family 34 (type II sodium/phosphate cotransporter), member 3; solute carrier family 34 member 3; type IIc Na+/Pi cotransporter; type IIc Na+/Pi-cotransporter; type IIc NaPi-cotransporter-v1
Gene Symbols SLC34A3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-117 of human SLC34A3 (NP_0639492).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 142680, 142681, 246234
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.