Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144223
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144223 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144223 Supplier Invitrogen™ Supplier No. PA5144223
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC35A1 Polyclonal Antibody for IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35A1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Slc35a1
Gene Accession No. P78382
Gene Alias AA408150; AI314851; CDG2F; CMP-SA-Tr; CMP-sialic acid transporter; CMP-Sia-Tr; CMPST; CST; hCST; mutated CMP-sialic acid transporter A1; Slc35a1; solute carrier family 35 (CMP-sialic acid transporter), member 1; solute carrier family 35 (CMP-sialic acid transporter), member A1; solute carrier family 35 (UDP-galactose transporter), member 1; solute carrier family 35 member A1
Gene Symbols Slc35a1
Host Species Rabbit
Immunogen Recombinant Human CMP-sialic acid transporter protein (1-100AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10559
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.