Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC35A2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA568493
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568493 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568493 Supplier Thermo Scientific Supplier No. PA568493

Rabbit Polyclonal Antibody

This gene encodes a member of the nucleotide-sugar transporter family. The encoded protein is a multi-pass membrane protein. It transports UDP-galactose from the cytosol into Golgi vesicles, where it serves as a glycosyl donor for the generation of glycans. Mutations in this gene cause congenital disorder of glycosylation type IIm (CDG2M). Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35A2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 23% sucrose and 0.09% sodium azide
Gene SLC35A2
Gene Accession No. P78381
Gene Alias UGALT, UGAT, UGT, UGT1, UGT2, UGTL, UDP-Gal-Tr, UDP-galactose translocator, UDP-galactose transporter, solute carrier family 35 (UDP-galactose transporter), member 2, solute carrier family 35 member A2
Gene Symbols SLC35A2
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human SLC35A2
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7355
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.