Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC35A4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566468
Description
Immunogen sequence: LLVGWQAWPQG PPPWRQAAPF ALS Highest antigen sequence identity to the following orthologs - mouse 88%, rat 92%.
SLC35A4 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane; positive regulation of translation in response to stress.
Specifications
SLC35A4 | |
Polyclonal | |
Unconjugated | |
SLC35A4 | |
Probable UDP-sugar transporter protein SLC35A4; SLC35A4; solute carrier family 35 (UDP-galactose transporter), member A4; solute carrier family 35 member A4; solute carrier family 35, member A4; tumor rejection antigen | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
113829 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96G79 | |
SLC35A4 | |
Recombinant Human SLC35A4. Recombinant protein control fragment (Product #RP-105226). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction