Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569284 Supplier Invitrogen™ Supplier No. PA569284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Guinea Pig: 85%; Horse: 87%; Human: 100%; Rabbit: 86%; Yeast: 100%.

SLC35A4 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane; positive regulation of translation in response to stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35A4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC35A4
Gene Accession No. Q96G79
Gene Alias Probable UDP-sugar transporter protein SLC35A4; SLC35A4; solute carrier family 35 (UDP-galactose transporter), member A4; solute carrier family 35 member A4; solute carrier family 35, member A4; tumor rejection antigen
Gene Symbols SLC35A4
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SLC35A4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113829
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.