Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557306
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557306 Supplier Invitrogen™ Supplier No. PA557306
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLQHNLAGRG FHHDAFFSPS NSCLLFRSEC PRKDNCTAKE WTFPEAKWNT TARV Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 69%.

This gene encodes a transmembrane protein which belongs to subfamily 35A of the solute carrier superfamily. The encoded protein is a nucleoside-sugar transporter. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35A5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC35A5
Gene Accession No. Q9BS91
Gene Alias probable UDP-sugar transporter protein SLC35A5; SLC35A5; Solute carrier family 35 member A5; solute carrier family 35, member A5; UNQ164/PRO190
Gene Symbols SLC35A5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC35A5. Recombinant protein control fragment (Product #RP-96343).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55032
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.