Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC35A5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566117
Description
Immunogen sequence: KPQVPEYAPRQ ERIRDLSGNL WERSSGDGEE LERLTKPKSD ESDEDTF Highest antigen sequence identity to the following orthologs - mouse 79%, rat 83%.
This gene encodes a transmembrane protein which belongs to subfamily 35A of the solute carrier superfamily. The encoded protein is a nucleoside-sugar transporter. Alternative splicing results in multiple transcript variants.
Specifications
SLC35A5 | |
Polyclonal | |
Unconjugated | |
SLC35A5 | |
probable UDP-sugar transporter protein SLC35A5; SLC35A5; Solute carrier family 35 member A5; solute carrier family 35, member A5; UNQ164/PRO190 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55032 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BS91 | |
SLC35A5 | |
Recombinant Human SLC35A5. Recombinant protein control fragment (Product #RP-106805). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction