Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565523
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565523 Supplier Invitrogen™ Supplier No. PA565523
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QFVNYPTQVLG KSCKPIPVML LGVTLLKKKY P Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

SLC35B1 belongs to the nucleotide-sugar transporter family and it is probable sugar transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35B1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC35B1
Gene Accession No. P78383
Gene Alias Endoplasmic reticulum nucleotide sugar transporter 1; ERNST; galactose transporter; hUGTrel1; putative endoplasmic reticulum nucleotide sugar transporter; Slc35b1; Solute carrier family 35 member B1; solute carrier family 35 member B1-like protein; solute carrier family 35, member B1; UDP-galactose translocator 2; UDP-galactose transporter related; UDP-galactose transporter-related protein 1; Ugalt2; UGALT2 protein; UGTREL1; wu:fj99g11; zgc:92284
Gene Symbols SLC35B1
Host Species Rabbit
Immunogen Recombinant Human SLC35B1. Recombinant protein control fragment (Product #RP-100118).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10237
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.