Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542450
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542450 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542450 Supplier Invitrogen™ Supplier No. PA542450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ALGQSFIFMT VVYFGPLTCS IITTTRKFFT ILASVILFAN PISPMQWVGT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

SLC35B1 belongs to the nucleotide-sugar transporter family and it is probable sugar transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35B1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC35B1
Gene Accession No. P78383
Gene Alias Endoplasmic reticulum nucleotide sugar transporter 1; ERNST; galactose transporter; hUGTrel1; putative endoplasmic reticulum nucleotide sugar transporter; Slc35b1; Solute carrier family 35 member B1; solute carrier family 35 member B1-like protein; solute carrier family 35, member B1; UDP-galactose translocator 2; UDP-galactose transporter related; UDP-galactose transporter-related protein 1; Ugalt2; UGALT2 protein; UGTREL1; wu:fj99g11; zgc:92284
Gene Symbols SLC35B1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SLC35B1 (aa 251-300).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10237
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.