Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523549
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523549 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523549 Supplier Invitrogen™ Supplier No. PA523549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC35B2 Polyclonal Antibody for Western Blot, IHC (P)

Mediates the transport of adenosine 3'-phospho 5'-phosphosulfate (PAPS), from cytosol into Golgi. PAPS is a universal sulfuryl donor for sulfation events that take place in the Golgi. May indirectly participate in activation of the NF-kappa-B and MAPK pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35B2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SLC35B2
Gene Accession No. Q8TB61
Gene Alias 1110003M08Rik; 3'-phosphoadenosine 5'-phosphosulfate transporter; AA407792; Adenosine 3'-phospho 5'-phosphosulfate transporter 1; AI842275; J207; PAPS transporter 1; PAPST1; PSEC0149; Putative MAPK-activating protein PM15; putative NF-kappa-B-activating protein 48; Slc35b1; SLC35B2; SLL; solute carrier family 35 (adenosine 3'-phospho 5'-phosphosulfate transporter), member B2; solute carrier family 35 member B2; solute carrier family 35 member B2 variant 2; solute carrier family 35, member B1; solute carrier family 35, member B2; UGTrel4
Gene Symbols SLC35B2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 393-421 amino acids from the C-terminal region of human S35B2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347734
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.