Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100883
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100883 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100883 Supplier Invitrogen™ Supplier No. PA5100883
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC35B2.

Mediates the transport of adenosine 3'-phospho 5'-phosphosulfate (PAPS), from cytosol into Golgi. PAPS is a universal sulfuryl donor for sulfation events that take place in the Golgi. May indirectly participate in activation of the NF-kappa-B and MAPK pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35B2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC35B2
Gene Accession No. Q8TB61, Q91ZN5
Gene Alias 1110003M08Rik; 3'-phosphoadenosine 5'-phosphosulfate transporter; AA407792; Adenosine 3'-phospho 5'-phosphosulfate transporter 1; AI842275; J207; PAPS transporter 1; PAPST1; PSEC0149; Putative MAPK-activating protein PM15; putative NF-kappa-B-activating protein 48; Slc35b1; SLC35B2; SLL; solute carrier family 35 (adenosine 3'-phospho 5'-phosphosulfate transporter), member B2; solute carrier family 35 member B2; solute carrier family 35 member B2 variant 2; solute carrier family 35, member B1; solute carrier family 35, member B2; UGTrel4
Gene Symbols SLC35B2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC35B2(Accession Q8TB61), corresponding to amino acid residues H388-V432.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347734, 501103, 73836
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.