Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC35B4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562043
Description
Immunogen sequence: GIFQETLYKR FGKHSKEALF Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

Specifications
SLC35B4 | |
Polyclonal | |
Unconjugated | |
SLC35B4 | |
PSEC0055; SLC35B4; solute carrier family 35 (UDP-xylose/UDP-N-acetylglucosamine transporter), member B4; solute carrier family 35 member B4; solute carrier family 35, member B4; UDP-xylose and UDP-N-acetylglucosamine transporter; UDP-Xylose/N-Acetylglucosamine transporter; YEA; YEA4; YEA4 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84912 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969S0 | |
SLC35B4 | |
Recombinant protein corresponding to Human SLC35B4. Recombinant protein control fragment (Product #RP-100317). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction