Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35C1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564146
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564146 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564146 Supplier Invitrogen™ Supplier No. PA564146
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WVRGWEMKKT PEEPSPKDSE KSAMGV Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.

SLC35C1 is involved in GDP-fucose import from the cytoplasm into the Golgi lumen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35C1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC35C1
Gene Accession No. Q96A29
Gene Alias CDG2C; E430007K15Rik; fuct1; GDP-fucose transporter; GDP-fucose transporter 1; GDP-Fuc-Tr; golgi GDP-fucose translocator; multispan transmembrane protein fuct1; putative golgi GDP-fucose translocator; SLC35C1; solute carrier family 35 (GDP-fucose transporter), member C1; solute carrier family 35 member C1; solute carrier family 35, member C1
Gene Symbols SLC35C1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC35C1. Recombinant protein control fragment (Product #RP-104180).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55343
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.