Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35C2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569273
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569273 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569273 Supplier Invitrogen™ Supplier No. PA569273
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 100%; Zebrafish: 100%.

May play an important role in the cellular response to tissue hypoxia. May be either a GDP-fucose transporter that competes with SLC35C1 for GDP-fucose, or a factor that otherwise enhances the fucosylation of Notch and is required for optimal Notch signaling in mammalian cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35C2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC35C2
Gene Accession No. Q8VCX2
Gene Alias BA394O2.1; C20orf5; C85957; CGI-15; D2Wsu58e; MGC137415 protein; ovarian cancer overexpressed 1; ovarian cancer overexpressed gene 1 protein; ovarian cancer-overexpressed gene 1 protein; Ovcov1; Slc35c2; slc35c2 protein; solute carrier family 35 (GDP-fucose transporter), member C2; solute carrier family 35 member C2; solute carrier family 35, member C2; wu:fb99e07; wu:fc49g11; wu:fi09e02; zgc:55285; zgc:77328
Gene Symbols SLC35C2
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of mouse SLC35C2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228875
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.