Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC35D1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP309534100UL

 View more versions of this product

Catalog No. NB123969


Only null left
Add to Cart

Description

Description

SLC35D1 Polyclonal specifically detects SLC35D1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SLC35D1
Polyclonal
Western Blot 1.0 ug/ml
KIAA0260MGC138236, solute carrier family 35 (UDP-glucuronic acid/UDP-N-acetylgalactosamine dualtransporter), member D1, Solute carrier family 35 member D1, UDP-galactose transporter-related protein 7, UDP-GlcA/UDP-GalNAc transporter, UDP-glucuronic acid/UDP-N-acetylgalactosamine transporter, UGTrel7, UGTREL7UDP-galactose transporter-related 7
The immunogen is a synthetic peptide directed towards the middle region of human SLC35D1 (NP_055954.1). Peptide sequence SDLAFDLEGYAFILINDVLTAANGAYVKQKLDSKELGKYGLLYYNALFM
100 μg
Primary
Human
Purified
Western Blot
Unconjugated
PBS buffer, 2% sucrose
Rabbit
Affinity purified
RUO
23169
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.