Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35D2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564148
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564148 Supplier Invitrogen™ Supplier No. PA564148
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YSFLTLSSQL KPKPVGEENI CLD Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 75%.

Antiporter transporting nucleotide sugars such as UDP-N-acetylglucosamine (UDP-GlcNAc), UDP-glucose (UDP-Glc) and GDP-mannose (GDP-Man) pooled in the cytosol into the lumen of the Golgi in exchange for the corresponding nucleosides monophosphates (UMP for UDP-sugars and GMP for GDP-sugars). May take part in heparan sulfate synthesis by supplying UDP-Glc-NAc, the donor substrate, and thus be involved in growth factor signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35D2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC35D2
Gene Accession No. Q76EJ3
Gene Alias 5730408I21Rik; fringe connection; hfrc; HFRC1; Homolog of Fringe connection protein 1; mUGTrel8; SLC35D2; solute carrier family 35 (UDP-GlcNAc/UDP-glucose transporter), member D2; solute carrier family 35 member D2; solute carrier family 35, member D2; SQV7L; SQV7-like protein; UDP-galactose transporter-related protein 8; UDP-glucuronic acid/UDP-N-acetylgalactosamine transporter; UDP-N-acetylglucosamine transporter; UDP-N-acetylglucosamine/UDP-glucose/GDP-mannose transporter; UGTrel8
Gene Symbols SLC35D2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC35D2. Recombinant protein control fragment (Product #RP-103541).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11046
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.