Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35D3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572721
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572721 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572721 Supplier Invitrogen™ Supplier No. PA572721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC35D3 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

May play a role in hemostasis as a regulator of the biosynthesis of platelet-dense granules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35D3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SLC35D3
Gene Accession No. Q5M8T2, Q8BGF8
Gene Alias 6230421J19Rik; frc, fringe-like 1; Frcl1; Fringe connection-like protein 1; SLC35D3; Solute carrier family 35 member D3; solute carrier family 35, member D3
Gene Symbols SLC35D3
Host Species Rabbit
Immunogen SLC35D3 antibody for a 17 amino acid synthetic peptide near the carboxy terminus of human SLC35D3. The immunogen is within the last 50 amino acids of SLC35D3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340146, 76157
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.