Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC35E1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297796MFV500
Description
SLC35E1 Polyclonal antibody specifically detects SLC35E1 in Human samples. It is validated for Western BlotSpecifications
SLC35E1 | |
Polyclonal | |
Western Blot | |
FLJ14251, FLJ36689, solute carrier family 35 member E1, solute carrier family 35, member E1 | |
Produced in rabbits immunized with E. coli-derived Human SLC35E1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
79939 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction