Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC35E1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566469
Description
Immunogen sequence: GPSPHPSSGPL LPPRFYPRYV LPL Highest antigen sequence identity to the following orthologs - mouse 92%, rat 96%.
Putative transporter.
Specifications
SLC35E1 | |
Polyclonal | |
Unconjugated | |
SLC35E1 | |
PSEC0038; SLC35E1; Solute carrier family 35 member E1; solute carrier family 35, member E1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79939 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96K37 | |
SLC35E1 | |
Recombinant Human SLC35E1. Recombinant protein control fragment (Product #RP-107144). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction