Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC35E2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564055
Description
Immunogen sequence: LGEYTGRPSD REEWEELQLQ PGRGAAASDR RSPVPPSERH GVRPHGENLP Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 34%.
Putative transporter.
Specifications
SLC35E2 | |
Polyclonal | |
Unconjugated | |
SLC35E2A | |
SLC35E2; SLC35E2A; solute carrier family 35 member E2; Solute carrier family 35 member E2A; solute carrier family 35, member E2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9906 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P0CK97 | |
SLC35E2A | |
Recombinant protein corresponding to Human SLC35E2B. Recombinant protein control fragment (Product #RP-101993). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction