Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35E2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561059 Supplier Invitrogen™ Supplier No. PA561059
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KARQHQQEAL QSLAAATGRA PDDTVEPLLP QDP Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 58%.

Putative transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35E2B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC35E2B
Gene Accession No. P0CK96
Gene Alias KIAA0447; SLC35E2; SLC35E2B; Solute carrier family 35 member E2B; solute carrier family 35, member E2B
Gene Symbols SLC35E2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC35E2B. Recombinant protein control fragment (Product #RP-99836).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728661
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.