Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC35F1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554029
Description
Immunogen sequence: RVYKQFRNPS GPVVDLPTTA QVEPSVTYTS LGQETEEEPH VRVA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 95%.
SLC35F1 is a putative solute transporter.
Specifications
SLC35F1 | |
Polyclonal | |
Unconjugated | |
SLC35F1 | |
C6orf169; dJ230I3.1; SLC35F1; Solute carrier family 35 member F1; solute carrier family 35, member F1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
222553 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T1Q4 | |
SLC35F1 | |
Recombinant protein corresponding to Human SLC35F1. Recombinant protein control fragment (Product #RP-91292). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction