Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC35F2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542470
Description
Peptide sequence: MEADSPAGPG APEPLAEGAA AEFSSLLRRI KGKLFTWNIL KTIALGQMLS Sequence homology: Human: 100%.
SLC35F2 is a putative solute transporter.
Specifications
SLC35F2 | |
Polyclonal | |
Unconjugated | |
SLC35F2 | |
1500009K05Rik; AU019213; HSNOV1; lung squamous cell cancer related protein LSCC-3; SLC35F2; Solute carrier family 35 member F2; solute carrier family 35, member F2 | |
Rabbit | |
Protein A | |
RUO | |
54733 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IXU6 | |
SLC35F2 | |
Synthetic peptide directed towards the N-terminal of human SLC35F2 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction