Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC35F2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565636
Description
Immunogen sequence: MEADSPAGPGA PEPLAEGAAA EFSSLLRRIK GKLFTWN Highest antigen sequence identity to the following orthologs - mouse 55%, rat 55%.
SLC35F2 is a putative solute transporter.
Specifications
SLC35F2 | |
Polyclonal | |
Unconjugated | |
SLC35F2 | |
1500009K05Rik; AU019213; HSNOV1; lung squamous cell cancer related protein LSCC-3; SLC35F2; Solute carrier family 35 member F2; solute carrier family 35, member F2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54733 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IXU6 | |
SLC35F2 | |
Recombinant Human SLC35F2. Recombinant protein control fragment (Product #RP-104913). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction