Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35F2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140229
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140229 Supplier Invitrogen™ Supplier No. PA5140229
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: MEADSPAGPG APEPLAEGAA AEFSSLLRRI KGKLFTWN Highest antigen sequence indentity to the following orthologs: Mouse - 55%, Rat - 55%.

SLC35F2 is a putative solute transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35F2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC35F2
Gene Accession No. Q8IXU6
Gene Alias 1500009K05Rik; AU019213; HSNOV1; lung squamous cell cancer related protein LSCC-3; SLC35F2; Solute carrier family 35 member F2; solute carrier family 35, member F2
Gene Symbols SLC35F2
Host Species Rabbit
Immunogen Recombinant protein fragment of Human SLC35F2 (Product #RP-104913)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54733
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.