Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC35F3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566405
Description
Immunogen sequence: WDVWLIKLLTR LKVRKKEEPA EGAADLSSGP QSKNRRARP Highest antigen sequence identity to the following orthologs - mouse 80%, rat 80%.
May be a thiamine transporter.
Specifications
SLC35F3 | |
Polyclonal | |
Unconjugated | |
SLC35F3 | |
B230375D17Rik; LOC100714042; Putative thiamine transporter SLC35F3; SLC35F3; Solute carrier family 35 member F3; solute carrier family 35, member F3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
148641 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IY50 | |
SLC35F3 | |
Recombinant Human SLC35F3. Recombinant protein control fragment (Product #RP-107082). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction