Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35F4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560538
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560538 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560538 Supplier Invitrogen™ Supplier No. PA560538
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEWDEITLRF INSLKEKKSE EHVDDVTDPS IHLRGRGRAN GTVSIPLA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.

Putative solute transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35F4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC35F4
Gene Accession No. A4IF30
Gene Alias 4930550L21Rik; c14_5373; C14orf36; MNCb-0335; RGD1561109; SLC35F4; Solute carrier family 35 member F4; solute carrier family 35, member F4
Gene Symbols SLC35F4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC35F4. Recombinant protein control fragment (Product #RP-89058).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 341880
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.