Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35F5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561912
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561912 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561912 Supplier Invitrogen™ Supplier No. PA561912
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLSSSPPFRL RSAKFSGIAL EDLRRALKTR LQMVCVFVMN RMNSQNSGFT QRR Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

SLC35F5 is a putative solute transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35F5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC35F5
Gene Accession No. Q8WV83
Gene Alias 1300003P13Rik; AI646727; HCV NS5A-transactivated protein 3; hepatitis C virus NS5A-transactivated protein 3; NS5ATP3; SLC35F5; Solute carrier family 35 member F5; solute carrier family 35, member F5; UNQ2545/PRO6097
Gene Symbols SLC35F5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC35F5. Recombinant protein control fragment (Product #RP-95999).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80255
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.