Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35F5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598692
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598692 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598692 Supplier Invitrogen™ Supplier No. PA598692
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC35F5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

SLC35F5 is a putative solute transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35F5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLC35F5
Gene Accession No. Q8WV83
Gene Alias 1300003P13Rik; AI646727; HCV NS5A-transactivated protein 3; hepatitis C virus NS5A-transactivated protein 3; NS5ATP3; SLC35F5; Solute carrier family 35 member F5; solute carrier family 35, member F5; UNQ2545/PRO6097
Gene Symbols SLC35F5
Host Species Rabbit
Immunogen Recombinant Human Solute carrier family 35 member F5 protein (122-242AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80255
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.