Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC36A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557472
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557472 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557472 Supplier Invitrogen™ Supplier No. PA557472
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSTQRLRNED YHDYSSTDVS PEESPSEGLN NLSSPGSYQR FGQSNSTTWF QT Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

This gene encodes a member of the eukaryote-specific amino acid/auxin permease (AAAP) 1 transporter family. The encoded protein functions as a proton-dependent, small amino acid transporter. This gene is clustered with related family members on chromosome 5q33.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC36A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC36A1
Gene Accession No. Q7Z2H8
Gene Alias Dct1; LYAAT1; lysosomal amino acid transporter 1; PAT1; proton/amino acid transporter 1; Proton-coupled amino acid transporter 1; SLC36A1; solute carrier family 36 (proton/amino acid symporter), member 1; solute carrier family 36 member 1; TRAMD3
Gene Symbols SLC36A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC36A1. Recombinant protein control fragment (Product #RP-96840).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 206358
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.