Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC36A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542529
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542529 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542529 Supplier Invitrogen™ Supplier No. PA542529
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TFLDESPSES AGLKKTKGIT VFQALIHLVK GNMGTGILGL PLAVKNAGIL Sequence homology: Human: 100%.

regulates pH-dependent, Na(+)-independent uptake of glycine, proline, and alpha(methyl)aminoisobutyric acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC36A2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC36A2
Gene Accession No. Q495M3
Gene Alias FLJ16051; MGC119658; MGC119660; PAT2; Proton/amino acid transporter 2; Proton-coupled amino acid transporter 2; SLC36A2; solute carrier family 36 (proton/amino acid symporter), member 2; solute carrier family 36 member 2; TRAMD1; tramdorin-1
Gene Symbols SLC36A2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC36A2 (aa 36-85).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153201
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.