Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC36A4 Antibody (PAT-4/9/H10), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250237AF750
Description
SLC36A4 Monoclonal specifically detects SLC36A4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Knockdown Validated.Specifications
SLC36A4 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
SLC36A4 | |
Antigenic amino acid sequence within the N-terminus of SLC36A4 (REELDMDVMRPLINE-C). | |
0.1 mL | |
Primary | |
Human | |
IgG2a κ |
Western Blot, Immunohistochemistry, KnockDown | |
PAT-4/9/H10 | |
Western Blot, Immunohistochemistry | |
proton/amino acid transporter 4, solute carrier family 36 (proton/amino acid symporter), member 4 | |
Mouse | |
Protein A purified | |
Cellular Signaling, metabolism | |
120103 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction