Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC36A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553658
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553658 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553658 Supplier Thermo Scientific Supplier No. PA553658
Add to Cart
Add to Cart

SLC36A4 Polyclonal Antibody

SLC36A4 Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC36A4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC36A4
Gene Accession No. Q6YBV0
Gene Alias 6330573I15Rik; PAT4; Proton/amino acid transporter 4; proton-coupled amino acid transporter 4; SLC36A4; solute carrier family 36 (proton/amino acid symporter), member 4; solute carrier family 36 member 4
Gene Symbols SLC36A4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC36A4. Recombinant protein control fragment (Product #RP-91238).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120103
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.