Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC36A4 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | SLC36A4 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4228
![]() ![]() |
Proteintech Group Inc
240661AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody can recognize the endogenous SLC36A4 protein in multiple cell lines.
SLC36A4 belongs to the SLC36 family of amino acid transporters based on sequence similarity with other family members (e.g., SLC36A1). SLC36 proteins contain about 500 amino acids and have 9 to 11 transmembrane domains. Unlike other SLC36 family members, which are proton-coupled amino acid transporters, SLC36A4 is a high-affinity/low-capacity non-proton-coupled amino acid transporter.Specifications
SLC36A4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q6YBV0 | |
120103 | |
SLC36A4 Fusion Protein Ag21142 | |
Primary | |
-20°C | |
SLC36A4 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
PAT4, SLC36A4 | |
SLC36A4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title