Learn More
Invitrogen™ SLC37A1 Polyclonal Antibody

Description
Immunogen sequence: IVKGELHKYC TAWDEADVRF SSQNRKSGSA APHQLPDNET DCGWAPFDKN NY Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 55%.
Specifications
Specifications
| Antigen | SLC37A1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SLC37A1 |
| Gene Accession No. | P57057 |
| Gene Alias | FLJ22340; G-3-P permease; G-3-P transporter; G3PP; Glucose-6-phosphate exchanger SLC37A1; glycerol-3-phosphate permease; glycerol-3-phosphate transporter; Slc37a1; solute carrier family 37 (glucose-6-phosphate transporter), member 1; solute carrier family 37 (glycerol-3-phosphate transporter), member 1; solute carrier family 37 member 1 |
| Gene Symbols | SLC37A1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.