Learn More
Invitrogen™ SLC37A2 Polyclonal Antibody

Description
A suggested positive control is mouse spleen tissue lysate. PA5-20916 can be used with blocking peptide PEP-1030.
Specifications
Specifications
| Antigen | SLC37A2 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SLC37A2 |
| Gene Accession No. | Q8TED4, Q9WU81 |
| Gene Alias | cAMP inducible 2 protein; cAMP inducible gene 2; cAMP-inducible protein 2; ci2; cI-2; G3PP; glucose-6-phosphate exchanger SLC37A2; pp11662; RGD1564160; Slc37a1; SLC37A2; solute carrier family 37 (glucose-6-phosphate transporter), member 2; solute carrier family 37 (glycerol-3-phosphate transporter), member 1; solute carrier family 37 (glycerol-3-phosphate transporter), member 2; solute carrier family 37 member 2; sugar phosphate exchanger 2 |
| Gene Symbols | SLC37A2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.