Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC37A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566428
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566428 Supplier Invitrogen™ Supplier No. PA566428
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VSPEEIGLSGI EAEENFEEDS HRPLINGGE Highest antigen sequence identity to the following orthologs - mouse 73%, rat 77%.

SLC37A3 (Solute Carrier Family 37 Member 3) is a Protein Coding gene. Diseases associated with SLC37A3 include Glycerol Kinase Deficiency and Deafness, Autosomal Recessive 8. Gene Ontology (GO) annotations related to this gene include transporter activity and transmembrane transporter activity. An important paralog of this gene is SLC37A1. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC37A3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC37A3
Gene Accession No. Q8NCC5
Gene Alias SLC37A3; solute carrier family 37 (glycerol-3-phosphate transporter), member 3; solute carrier family 37 member 3; solute carrier family 37, member 3; SPX3; Sugar phosphate exchanger 3; sugar phosphate exchanger 3-like protein
Gene Symbols SLC37A3
Host Species Rabbit
Immunogen Recombinant Human SLC37A3. Recombinant protein control fragment (Product #RP-105213).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84255
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.