Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC37A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542500
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542500 Supplier Invitrogen™ Supplier No. PA542500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FFGLLVSPEE IGLSGIEAEE NFEEDSHRPL INGGENEDEY EPNYSIQDDS Sequence homology: Cow: 92%; Dog: 86%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Rabbit: 93%; Rat: 85%.

SLC37A3 (Solute Carrier Family 37 Member 3) is a Protein Coding gene. Diseases associated with SLC37A3 include Glycerol Kinase Deficiency and Deafness, Autosomal Recessive 8. Gene Ontology (GO) annotations related to this gene include transporter activity and transmembrane transporter activity. An important paralog of this gene is SLC37A1. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC37A3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC37A3
Gene Accession No. Q8NCC5
Gene Alias SLC37A3; solute carrier family 37 (glycerol-3-phosphate transporter), member 3; solute carrier family 37 member 3; solute carrier family 37, member 3; SPX3; Sugar phosphate exchanger 3; sugar phosphate exchanger 3-like protein
Gene Symbols SLC37A3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SLC37A3.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84255
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.