Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC37A4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542422
Description
Peptide sequence: LVEEIPLDKD DLGFITSSQS AAYAISKFVS GVLSDQMSAR WLFSSGLLLV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 93%; Rat: 93%.
SLC37A4 transports glucose-6-phosphate from the cytoplasm to the lumen of the endoplasmic reticulum. It forms with glucose-6-phosphatase the complex responsible for glucose production through glycogenolysis and gluconeogenesis. Hence, it plays a central role in homeostatic regulation of blood glucose levels.
Specifications
SLC37A4 | |
Polyclonal | |
Unconjugated | |
Slc37a4 | |
G6PT; G6PT1; G6PT2; G6PT3; glucose 6-phosphate transporter; glucose-5-phosphate transporter; glucose-6-phosphatase transport protein 1; glucose-6-phosphatase, transport (glucose) protein 3; glucose-6-phosphatase, transport (glucose-6-phosphate) protein 1; glucose-6-phosphatase, transport (phosphate/pyrophosphate) protein 2; glucose-6-phosphatase, transport protein 1; glucose-6-phosphate exchanger SLC37A4; Glucose-6-phosphate translocase; GSD1b; GSD-1b; GSD1c; GSD1d; mG6PT; MGC15729; microsomal glucose-6-phosphate transporter; PRO0685; SLC37A4; solute carrier family 37 (glucose-6-phosphate transporter), member 4; solute carrier family 37 (glycerol-6-phosphate transporter), member 4; solute carrier family 37 member 4; Transformation-related gene 19 protein; transporter; TRG19; TRG-19; Unknown (protein for MGC:137413) | |
Rabbit | |
Affinity chromatography | |
RUO | |
2542 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O43826 | |
Slc37a4 | |
Synthetic peptide directed towards the N-terminal of human SLC37A4 (aa 38-87). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction