Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC37A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558600
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558600 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558600 Supplier Invitrogen™ Supplier No. PA558600
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NEPADVGLRN LDPMPSEGKK GSLKEESTLQ E Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

SLC37A4 transports glucose-6-phosphate from the cytoplasm to the lumen of the endoplasmic reticulum. It forms with glucose-6-phosphatase the complex responsible for glucose production through glycogenolysis and gluconeogenesis. Hence, it plays a central role in homeostatic regulation of blood glucose levels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC37A4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc37a4
Gene Accession No. O43826
Gene Alias G6PT; G6PT1; G6PT2; G6PT3; glucose 6-phosphate transporter; glucose-5-phosphate transporter; glucose-6-phosphatase transport protein 1; glucose-6-phosphatase, transport (glucose) protein 3; glucose-6-phosphatase, transport (glucose-6-phosphate) protein 1; glucose-6-phosphatase, transport (phosphate/pyrophosphate) protein 2; glucose-6-phosphatase, transport protein 1; glucose-6-phosphate exchanger SLC37A4; Glucose-6-phosphate translocase; GSD1b; GSD-1b; GSD1c; GSD1d; mG6PT; MGC15729; microsomal glucose-6-phosphate transporter; PRO0685; SLC37A4; solute carrier family 37 (glucose-6-phosphate transporter), member 4; solute carrier family 37 (glycerol-6-phosphate transporter), member 4; solute carrier family 37 member 4; Transformation-related gene 19 protein; transporter; TRG19; TRG-19; Unknown (protein for MGC:137413)
Gene Symbols Slc37a4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC37A4. Recombinant protein control fragment (Product #RP-95794).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2542
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.