Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC37A4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985854
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5854 Supplier Bioss Supplier No. BS4039R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC37A4 transports glucose-6-phosphate from the cytoplasm to the lumen of the endoplasmic reticulum. It forms with glucose-6-phosphatase the complex responsible for glucose production through glycogenolysis and gluconeogenesis. Hence, it plays a central role in homeostatic regulation of blood glucose levels.

Specifications

Antigen SLC37A4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Slc37a4
Gene Accession No. O43826
Gene Alias BOS_14605; G6PT; G6PT1; G6PT2; G6PT3; glucose 6-phosphate transporter; Glucose-5-phosphate transporter; glucose-6-phosphatase transport protein 1; glucose-6-phosphatase, transport (glucose) protein 3; glucose-6-phosphatase, transport (glucose-6-phosphate) protein 1; glucose-6-phosphatase, transport (phosphate/pyrophosphate) protein 2; glucose-6-phosphatase, transport protein 1; Glucose-6-phosphate exchanger SLC37A4; Glucose-6-phosphate translocase; GSD1b; GSD-1b; GSD1c; GSD1d; mG6PT; MGC15729; microsomal glucose-6-phosphate transporter; PRO0685; SLC37A4; solute carrier family 37 (glucose-6-phosphate transporter), member 4; solute carrier family 37 (glycerol-6-phosphate transporter), member 4; Solute carrier family 37 member 4; Transformation-related gene 19 protein; transporter; TRG19; TRG-19
Gene Symbols Slc37a4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SLC37A4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2542
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.