Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC38A1 Antibody (S104-32), Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | SLC38A1 |
---|---|
Clone | S104-32 |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
SLC38A1 Monoclonal specifically detects SLC38A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
SLC38A1 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
81539 | |
Fusion protein amino acids 1- 63 of rat SNAT1 | |
Primary | |
Detects 50kDa. |
S104-32 | |
Monoclonal | |
Purified | |
RUO | |
Amino acid transporter A1, ATA1SNAT1, NAT2N-system amino acid transporter 2, SAT1amino acid transporter system A1, sodium-coupled neutral amino acid transporter 1, Solute carrier family 38 member 1, solute carrier family 38, member 1, System A amino acid transporter 1, System N amino acid transporter 1 | |
SLC38A1 | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title