Learn More
Invitrogen™ SLC38A1 Monoclonal Antibody (N104/32), PE
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA545532
Description
1 μg/mL of MA5-45532 was sufficient for detection of SNAT1 in 20 μg of lysates from neocortical neurons cultured under amino acid starvation conditions and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 50kDa. This antibody was formerly sold as clone S104-32.
Amino acid transporters play essential roles in the uptake of nutrients, production of energy, chemical metabolism, detoxification, and neurotransmitter cycling. SLC38A1 is an important transporter of glutamine, an intermediate in the detoxification of ammonia and the production of urea. Glutamine serves as a precursor for the synaptic transmitter, glutamate.
Specifications
SLC38A1 | |
Monoclonal | |
1 mg/mL | |
2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
Q8K2P7, Q9H2H9, Q9JM15 | |
SLC38A1 | |
Fusion protein amino acids 1- 63 of rat SNAT1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
N104/32 | |
PE | |
SLC38A1 | |
AA408026; AA409865; AL022800; Amino acid transporter A1; amino acid transporter system A1; ATA1; AU015942; GlnT; glutamine transporter; MNat2; Nat2; N-system amino acid transporter 2; rATA1; S38A1; Sa2; SAT1; SLC38A1; SNAT 1; Snat1; sodium-coupled neutral amino acid transporter 1; Solute carrier family 38 member 1; solute carrier family 38, member 1; system A amino acid transporter 1; System A transporter 2; system N amino acid transporter 1 | |
Mouse | |
Protein G | |
RUO | |
105727, 170567, 81539 | |
4°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.