Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC38A1 Monoclonal Antibody (N104/32)
GREENER_CHOICE

Catalog No. PIMA527605
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527605 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527605 Supplier Invitrogen™ Supplier No. MA527605
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

1 μg/mL of MA5-27605 was sufficient for detection of SNAT1 in 20 μg of lysates from neocortical neurons cultured under amino acid starvation conditions and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 50kDa. This antibody was formerly sold as clone S104-32.

Amino acid transporters play essential roles in the uptake of nutrients, production of energy, chemical metabolism, detoxification, and neurotransmitter cycling. SLC38A1 is an important transporter of glutamine, an intermediate in the detoxification of ammonia and the production of urea. Glutamine serves as a precursor for the synaptic transmitter, glutamate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC38A1
Applications Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N104/32
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene SLC38A1
Gene Accession No. Q8K2P7, Q9H2H9, Q9JM15
Gene Alias AA408026; AA409865; AL022800; Amino acid transporter A1; amino acid transporter system A1; ATA1; AU015942; GlnT; glutamine transporter; MNat2; Nat2; N-system amino acid transporter 2; rATA1; S38A1; Sa2; SAT1; SLC38A1; SNAT 1; Snat1; sodium-coupled neutral amino acid transporter 1; Solute carrier family 38 member 1; solute carrier family 38, member 1; system A amino acid transporter 1; System A transporter 2; system N amino acid transporter 1
Gene Symbols SLC38A1
Host Species Mouse
Immunogen Fusion protein amino acids 1-63 of rat SNAT1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105727, 170567, 81539
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.