Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC38A1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984378
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4378 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4378 Supplier Bioss Supplier No. BS19825R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Amino acid transporters play essential roles in the uptake of nutrients, production of energy, chemical metabolism, detoxification, and neurotransmitter cycling. SLC38A1 is an important transporter of glutamine, an intermediate in the detoxification of ammonia and the production of urea. Glutamine serves as a precursor for the synaptic transmitter, glutamate.

Specifications

Antigen SLC38A1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLC38A1
Gene Accession No. Q9H2H9
Gene Alias AA408026; AA409865; AL022800; Amino acid transporter A1; amino acid transporter system A1; Ata1; AU015942; BOS_5172; Glnt; Glutamine transporter; LOC100357643; MNat2; Nat2; N-system amino acid transporter 2; rATA1; S38A1; Sa2; Sat1; SLC38A1; SNAT 1; Snat1; Sodium-coupled neutral amino acid transporter 1; Solute carrier family 38 member 1; solute carrier family 38, member 1; solute carrier family 38, member 1-like; System A amino acid transporter 1; System A transporter 2; System N amino acid transporter 1
Gene Symbols SLC38A1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SLC38A1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81539
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.